
Camostat methanesulphate ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:59721-29-8
:
Formula:C₂₀H₂₂N₄O₅·CH₃SO₃H
MDL Number:MFCD00941410
:494.52 g/mol
Merck Index:14,01728
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Nitrogen):11.10 to 11.60 %
- Melting point:196.0 to 200.0 deg-C
Specification Test Results
Frequently Bought Together