Order Entry
Netherlands
ContactUsLinkComponent
1-(4-Chlorophenyl)-2-thiourea ≥98.0% (by HPLC, total nitrogen)
Toxic
1-(4-Chlorophenyl)-2-thiourea ≥98.0% (by HPLC, total nitrogen)
Supplier:  TCI
Certificates
Toxic
1-(4-Chlorophenyl)-2-thiourea ≥98.0% (by HPLC, total nitrogen)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Melting point:
    169.0 to 173.0 deg-C

Specification Test Results

Frequently Bought Together