Order Entry
Netherlands
ContactUsLinkComponent
Cyclopentyl isocyanide ≥98.0% (by GC)
Irritant
Cyclopentyl isocyanide ≥98.0% (by GC)
Catalog # TCIAC2959-1G
Supplier:  TCI
CAS Number:  
Irritant
Cyclopentyl isocyanide ≥98.0% (by GC)
Catalog # TCIAC2959-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
Boiling Pt:28°C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:68498-54-4
Formula:C₆H₉N
MDL Number:MFCD02664624