
2-Chloro-6-methylquinoline-3-carbaldehyde ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:73568-27-1
:
Formula:C₁₁H₈ClNO
MDL Number:MFCD01912773
:205.6400 g/mol
:GHS07
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (GC):min. 96.0 %
- Melting point:122.0 to 126.0 deg-C
Specification Test Results
Frequently Bought Together