
6-[(3-Chloropropyl)amino]-1,3-dimethyluracil ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAC2929-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:156.0 to 160.0 deg-C
Specification Test Results
About this item
:
:231.68 g/mol
EINECS:252-129-9
:GHS07
CAS Number:34654-81-4
Formula:C₉H₁₄ClN₃O₂
MDL Number:MFCD00453355