
4-Chloro-3-fluorobenzoic acid ≥98.0% (by GC, titration analysis)
Catalog # TCIAC2891-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:193.0 to 197.0 deg-C
Specification Test Results
About this item
:
:174.56 g/mol
:GHS07
CAS Number:403-17-8
Formula:C₇H₄ClFO₂
MDL Number:MFCD00143290