Order Entry
Netherlands
ContactUsLinkComponent
4-Chloro-2,3-dimethylpyridine-N-oxide ≥98.0% (by GC, titration analysis)
Irritant
Health
4-Chloro-2,3-dimethylpyridine-N-oxide ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
Irritant
Health
4-Chloro-2,3-dimethylpyridine-N-oxide ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    106.0 to 110.0 deg-C

Specification Test Results

Frequently Bought Together