
5-(2-Chlorophenyl)-1H-tetrazole ≥98.0% (by HPLC, titration analysis)
: TCI
:50907-46-5
:
:C₇H₅ClN₄
:MFCD00040950
:180.6 g/mol
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
Frequently Bought Together