
Catecholbis(trifluoromethanesulphonate) ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:17763-91-6
:
Formula:C₈H₄F₆O₆S₂
MDL Number:MFCD00274274
:374.22 g/mol
UN:3261
ADR:8,II
:GHS05
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:35.0 to 39.0 deg-C
Specification Test Results
Frequently Bought Together