Order Entry
Netherlands
ContactUsLinkComponent
4-Chlorobenzofuro[3,2-d]pyrimidine ≥98.0% (by GC)
4-Chlorobenzofuro[3,2-d]pyrimidine ≥98.0% (by GC)
Catalog # TCIAC2800-200MG
Supplier:  TCI
CAS Number:  
4-Chlorobenzofuro[3,2-d]pyrimidine ≥98.0% (by GC)
Catalog # TCIAC2800-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    White to Yellow to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    144.0 to 148.0 deg-C

Specification Test Results

About this item

 :
 :204.6100 g/mol
CAS Number:39876-88-5
Formula:C₁₀H₅ClN₂O
MDL Number:MFCD00763943