Order Entry
Netherlands
ContactUsLinkComponent
Calcium dobesilate hydrate ≥98.0% (by HPLC)
 
Calcium dobesilate hydrate ≥98.0% (by HPLC)
  TCIAC2789-1G
 :  TCI
 :  
 
Calcium dobesilate hydrate ≥98.0% (by HPLC)
  TCIAC2789-1G
 :  TCI
 :  

 

  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Chelometric Titration):
    min. 95.0 % (calcd.on anh.substance)
  • Water:
    3.0 to 7.0 %

 

 

 :
 :418.40 g/mol
 :14,03394
 :243-531-5
 :GHS07
 :117552-78-0
 :C₆H₆O₅S
 :MFCD00867249