Order Entry
Netherlands
ContactUsLinkComponent
S-(2-Carboxyethyl)-L-cysteine ≥98.0% (by titrimetric analysis)
S-(2-Carboxyethyl)-L-cysteine ≥98.0% (by titrimetric analysis)
Catalog # TCIAC2781-100MG
Supplier:  TCI
CAS Number:  
S-(2-Carboxyethyl)-L-cysteine ≥98.0% (by titrimetric analysis)
Catalog # TCIAC2781-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Specific rotation [a]20/D:
    -8.0 to -11.0 deg (C=2,1mol/LHCl)

Specification Test Results

About this item

 :
 :193.22 g/mol
EINECS:223-717-2
CAS Number:4033-46-9
Formula:C₆H₁₁NO₄S
MDL Number:MFCD03424208