

4-Chloro-3-(trifluoromethyl)phenyl isocyanate ≥98.0% (by GC)
: TCI
:327-78-6
:
:C₈H₃ClF₃NO
:MFCD00013874
:221.57 g/mol
:92°C
:2206
:6.1,III
:GHS06
:GHS08
:Refrigerator
- Appearance:White to Almost white powder to lump
- Purity (GC):min. 98.0 %
- Melting point:34.0 to 38.0 deg-C
Frequently Bought Together