
2-Chloro-4-fluorophenylboronic acid (contains varying amounts of Anhydride)
Catalog # TCIAC2772-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):97.0 to 112.0 %
Specification Test Results
About this item
:
:174.37 g/mol
Melting Pt:237°C
:GHS07
CAS Number:313545-72-1
Formula:C₆H₅BClFO₂
MDL Number:MFCD02684295