
3-Chloro-4-fluorobenzylamine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:72235-56-4
:
Formula:C₇H₇ClFN
MDL Number:MFCD00042507
:159.59 g/mol
UN:2735
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together