
ɑ-Cyano-3-hydroxycinnamic acid ≥98.0% (by HPLC, titration analysis)
TCIAC2677-5G
: TCI
:
- Pk:5 g
- Seq:3173
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
:
:189.17 g/mol
:259-289-9
:GHS07
:54673-07-3
:C₁₀H₇NO₃
:MFCD00004203