Order Entry
Netherlands
ContactUsLinkComponent
3-Chloro-2-fluoro-5-(trifluoromethyl)pyridine ≥98.0% (by GC)
Flammable
Irritant
3-Chloro-2-fluoro-5-(trifluoromethyl)pyridine ≥98.0% (by GC)
Catalog # TCIAC2671-5G
Supplier:  TCI
CAS Number:  
Flammable
Irritant
3-Chloro-2-fluoro-5-(trifluoromethyl)pyridine ≥98.0% (by GC)
Catalog # TCIAC2671-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
Flash Pt:52°C
UN:1993
ADR:3,III
 :199.53 g/mol
 :GHS02
 :GHS07
CAS Number:72537-17-8
Formula:C₆H₂ClF₄N
MDL Number:MFCD00191918