Ciprofibrate ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:52214-84-3
:
Formula:C₁₃H₁₄Cl₂O₃
MDL Number:MFCD00467135
:289.15 g/mol
Merck Index:14,02313
Storage Temperature:Refrigerator
EINECS:257-744-6
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:116.0 to 120.0 deg-C
Specification Test Results
Frequently Bought Together