Order Entry
Netherlands
ContactUsLinkComponent
 
Chitotriose trihydrochloride hydrate ≥93.0% (by HPLC)
Catalog # TCIAC2642-25MG
Supplier:  TCI
CAS Number:  
Chitotriose trihydrochloride hydrate ≥93.0% (by HPLC)
Catalog # TCIAC2642-25MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 93.0 area%
  • Elemental analysis (Nitrogen):
    5.8 to 7.2 %
  • Specific rotation [a]20/D:
    +26.0 to +31.0 deg (C=1, H2O) (calcd.on anh.substance)
  • Water:
    max. 10.0 %

Specification Test Results

About this item

 :
 :610.86 g/mol
Melting Pt:255°C
CAS Number:117436-78-9
Formula:C₁₈H₃₅N₃O₁₃