Chitotetraose tetrahydrochloride hydrate ≥95.0% (by HPLC)
TCIAC2641-25MG
: TCI
:
- Pk:25 mg
- Seq:2315
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 95.0 area%
- Elemental analysis (Nitrogen):5.9 to 7.2 %
- Specific rotation [a]20/D:+18.0 to +21.0 deg (C=1、H2O) (calcd.on anh.substance)
- Water:max. 12.0 %
:
:808.48 g/mol
:117399-50-5
:C₂₄H₄₆N₄O₁₇