Copper(I) hexafluoro-2,4-pentanedionate-1,5-cyclooctadiene complex ≥95.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:86233-74-1
:
Formula:C₁₃H₁₃CuF₆O₂
MDL Number:MFCD00156517
:378.78 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Light yellow to Yellow to Green powder to crystal
- Purity (Chelometric Titration):min. 95.0 %
Specification Test Results
Frequently Bought Together