Order Entry
Netherlands
ContactUsLinkComponent
4-(3-Chloropropyl)morpholine ≥98.0% (by GC, titration analysis)
Irritant
4-(3-Chloropropyl)morpholine ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
Irritant
4-(3-Chloropropyl)morpholine ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

Frequently Bought Together