
N-Cbz-4-piperidinecarboxylic acid ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:10314-98-4
:
Formula:C₁₄H₁₇NO₄
MDL Number:MFCD01568759
:263.2900 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Neutralization titration):min. 97.0 %
- Melting point:71.0 to 75.0 deg-C
Specification Test Results
Frequently Bought Together