
1,1-Cyclohexanediacetic anhydride ≥98.0% (by GC)
Catalog # TCIAC2533-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Morpholine Method):min. 96.0 %
- Melting point:68.0 to 72.0 deg-C
Specification Test Results
About this item
:
:182.22 g/mol
Boiling Pt:126°C
:GHS07
CAS Number:1010-26-0
Formula:C₁₀H₁₄O₃
MDL Number:MFCD00040292