Chloro[(1,3-dimesitylimidazol-2-ylidene)(N,N-dimethylbenzylamine)palladium(II)] ≥98.0% (by titrimetric analysis)
: TCI
:1058661-78-1
:
:C₃₀H₃₆ClN₃Pd
:580.51 g/mol
- Appearance:White to Almost white powder to crystal
- Purity (Argentmetric Titration):min. 98.0 %
- Purity (Chelometric back titration):min. 98.0 %
Frequently Bought Together