Order Entry
Netherlands
ContactUsLinkComponent
N-Carbobenzoxy-L-valine succinimidyl ester ≥98.0% (by HPLC)
N-Carbobenzoxy-L-valine succinimidyl ester ≥98.0% (by HPLC)
Catalog # TCIAC2334-5G
Supplier:  TCI
CAS Number:  
N-Carbobenzoxy-L-valine succinimidyl ester ≥98.0% (by HPLC)
Catalog # TCIAC2334-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Melting point:
    114.0 to 118.0 deg-C
  • Specific rotation [a]20/D:
    -23.0 to -26.0 deg (C=1, Dioxane)

Specification Test Results

About this item

 :
 :348.36 g/mol
Storage Temperature:Freezer
EINECS:222-493-3
CAS Number:3496-11-5
Formula:C₁₇H₂₀N₂O₆
MDL Number:MFCD00053547