
Cetirizine dihydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:83881-52-1
:
Formula:C₂₁H₂₅ClN₂O₃
MDL Number:MFCD00941428
:461.81 g/mol
Merck Index:14,02022
Melting Pt:225°C
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together