Coumarin 102 ≥97.0% (by HPLC, total nitrogen)
Catalog # TCIAC2267-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (with Total Nitrogen):min. 97.0 %
- Melting point:149.0 to 153.0 deg-C
Specification Test Results
About this item
:
:255.32 g/mol
EINECS:255-285-6
CAS Number:41267-76-9
Formula:C₁₆H₁₇NO₂
MDL Number:MFCD00041844