1,4-Chrysenequinone ≥93.0% (by HPLC)
Catalog # TCIAC2214-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 93.0 area%
- Melting point:211.0 to 217.0 deg-C
Specification Test Results
About this item
:
:258.28 g/mol
CAS Number:100900-16-1
Formula:C₁₈H₁₀O₂
MDL Number:MFCD00009941