
D-Cysteine methyl ester hydrochloride ≥98.0% (by titrimetric analysis)
: TCI
:70361-61-4
:MFCD00672724
:C₄H₉NO₂S
:171.64 g/mol
:GHS07
- Appearance:White to Almost white powder to crystaline
- Purity (Iodometric Titration):min. 98.0 %
- Specific rotation [a]20/D:+1.5 to +3.5 deg (C=20, MeOH)
- Solubility in Water:almost transparency
Frequently Bought Together