Order Entry
Netherlands
ContactUsLinkComponent
Carbon nanotube multi-walled, Length: 5-15 µm, Ø 10-20 nm
Irritant
Carbon nanotube multi-walled, Length: 5-15 µm, Ø 10-20 nm
Supplier:  TCI
Certificates
Irritant
Carbon nanotube multi-walled, Length: 5-15 µm, Ø 10-20 nm
Supplier:  TCI

Specification Test Results

  • Appearance:
    Black powder to crystal
  • Purity (TPO Method):
    min. 97 %
  • Diameter:
    10 to 20 nm
  • Length:
    5 to 15 micro_m

Specification Test Results

Related Information

Frequently Bought Together