Order Entry
Netherlands
ContactUsLinkComponent
 
 
1,3,5-Cyclohexanetricarboxylic acid (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
 :  TCI
 
 
1,3,5-Cyclohexanetricarboxylic acid (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 % (total of isomers)
  • Purity (Neutralization titration):
    min. 98.0 %

 

Frequently Bought Together