

1-Cyclohexyl-5-(4-chlorobutyl)-1H-tetrazole ≥98.0% (by HPLC)
TCIAC2013-5G
: TCI
:
- Pk:5 g
- Seq:3173
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:50.0 to 54.0 deg-C
:
:242.75 g/mol
:1325
:4.1,III
:GHS02
:GHS07
:73963-42-5
:C₁₁H₁₉ClN₄
:MFCD01076191