
1-(4-Carboxyphenyl)-5-mercapto-1H-tetrazole ≥95.0% (by HPLC, titration analysis)
Catalog # TCIAC2011-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (Neutralization titration):min. 95.0 %
Specification Test Results
About this item
:
:222.22 g/mol
UN:1325
ADR:4.1,II
:GHS02
CAS Number:23249-95-8
Formula:C₈H₆N₄O₂S
MDL Number:MFCD01074835