Order Entry
Netherlands
ContactUsLinkComponent
 
 
 
Chloromethyl cyclohexyl ether ≥96.0% (by GC)
  TCIAC1947-25G
 :  TCI
 :  
 
 
Chloromethyl cyclohexyl ether ≥96.0% (by GC)
  TCIAC1947-25G
 :  TCI
 :  

 

  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 96.0 %

 

 

 :
 :148.63 g/mol
 :185°C
 :GHS05
 :GHS07
 :3587-62-0
 :C₇H₁₃ClO
 :MFCD10566884