4-(4-Chlorophenyl)thiomorpholine-1,1-dioxide ≥98.0% (by GC, titration analysis)
TCIAC1935-5G
: TCI
:
- Pk:5 g
- Seq:3173
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
- Melting point:155.0 to 159.0 deg-C
- Solubility in hot Methanol:very faint turbidity
:
:245.72 g/mol
:82222-74-0
:C₁₀H₁₂ClNO₂S
:MFCD07366283