
cis-1,4-Cyclohexanediamine ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:15827-56-2
:
Formula:C₆H₁₄N₂
MDL Number:MFCD08276323
Boiling Pt:90°C
UN:2735
ADR:8,III
:114.19 g/mol
:GHS05
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 96.0 %
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together