Order Entry
Netherlands
ContactUsLinkComponent
Tetrachloro-p-benzoquinone ∼2% in DMF for detection of primary and secondary amines
Tetrachloro-p-benzoquinone ∼2% in DMF for detection of primary and secondary amines
Catalog # TCIAC1770-10ML
Supplier:  TCI
CAS Number:  
undefined
Tetrachloro-p-benzoquinone ∼2% in DMF for detection of primary and secondary amines
Catalog # TCIAC1770-10ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    10 mL
  • Seq:
    2275

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Brown clear liquid
  • Concentration (Iodometric titration):
    2.0 to 2.3 w/w%

Specification Test Results

About this item

 :
 :245.88 g/mol
Merck Index:14,02078
EINECS:204-274-4
UN:2265
ADR:3,III
 :GHS02
 :GHS05
 :GHS06
 :GHS08
CAS Number:118-75-2
Formula:C₆Cl₄O₂
MDL Number:MFCD00001594