N-(2-Carboxy-4,5-dichlorobenzoyl)-(+)-10,2-camphorsultam ≥98.0% (by HPLC, titration analysis)
TCIAC1767-500MG
: TCI
:
- Pk:500 mg
- Seq:2725
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Optical purity (LC):min. 98.0 ee%
- Purity (Neutralization titration):min. 98.0 %
- Specific rotation [a]20/D:+90.0 to +100.0 deg (C=1, MeOH)
:
:432.31 g/mol
:C₁₈H₁₉Cl₂NO₅S
:MFCD04117916