
Cyclobis(paraquat-1,4-phenylene)tetrakis(hexafluorophosphate) ≥94.0% (by HPLC)
Catalog # TCIAC1749-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Purity (HPLC):min. 94.0 area%
Specification Test Results
About this item
:
:1100.54 g/mol
Melting Pt:275°C
:GHS07
CAS Number:117271-77-9
Formula:C₃₆H₃₂F₂₄N₄P₄