
2-[4-(Cyanomethyl)phenyl]acetamide ≥97.0% (by total nitrogen basis)
Catalog # TCIAC1748-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (with Total Nitrogen):min. 97.0 %
- Melting point:163.0 to 167.0 deg-C
Specification Test Results
About this item
:
:174.20 g/mol
UN:3439
ADR:6.1,III
:GHS06
CAS Number:99071-55-3
Formula:C₁₀H₁₀N₂O