N-(2-Carboxy-4,5-dichlorobenzoyl)-(-)-10,2-camphorsultam ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAC1683-500MG
Supplier: TCI
CAS Number:
Specifications
- Pk:500 mg
- Seq:2725
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Optical purity (LC):min. 98.0 ee%
- Purity (Neutralization titration):min. 98.0 %
- Specific rotation [a]20/D:-90.0 to -100.0 deg (C=1, MeOH)
Specification Test Results
About this item
:
:432.31 g/mol
CAS Number:193202-37-8
Formula:C₁₈H₁₉Cl₂NO₅S
MDL Number:MFCD04117914