N-(2-Carboxybenzoyl)-(-)-10,2-camphorsultam ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAC1682-500MG
Supplier: TCI
CAS Number:
Specifications
- Pk:500 mg
- Seq:2725
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Optical purity (LC):min. 98.0 ee%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:181.0 to 185.0 deg-C
Specification Test Results
About this item
:
:363.43 g/mol
CAS Number:179950-32-4
Formula:C₁₈H₂₁NO₅S
MDL Number:MFCD04117913