Order Entry
Netherlands
ContactUsLinkComponent
2-Chloro-1,3-dimethylimidazolinium chloride ∼25% in dichloromethane
Irritant
Health
Environment
2-Chloro-1,3-dimethylimidazolinium chloride ∼25% in dichloromethane
Catalog # TCIAC1639-25G
Supplier:  TCI
CAS Number:  
Irritant
Health
Environment
2-Chloro-1,3-dimethylimidazolinium chloride ∼25% in dichloromethane
Catalog # TCIAC1639-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Brown clear liquid
  • Concentration (Argentmetric Titration):
    25.0 to 29.0 w/w%

Specification Test Results

About this item

 :
 :169.05 g/mol
Storage Temperature:Refrigerator
UN:1593
ADR:6.1,III
 :GHS07
 :GHS08
 :GHS09
CAS Number:37091-73-9
Formula:C₅H₁₀Cl₂N₂
MDL Number:MFCD00137463