
1-(3-Chlorophenyl)-4-(3-chloropropyl)piperazine hydrochloride ≥97.0% (by HPLC, titration analysis)
TCIAC1608-25G
: TCI
:
- Pk:25 g
- Seq:3308
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Nonaqueous Titration):min. 97.0 %
- Melting point:199.0 to 203.0 deg-C
- Solubility in Acetic acid:almost transparency
:
:309.66 g/mol
:258-038-0
:GHS07
:52605-52-4
:C₁₃H₁₈Cl₂N₂
:MFCD00067039