Ga naar Rubriek
Ga naar Hoofdinhoud
Ga naar Voettekst
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Chloromethyl-p-tolyl sulphide
Chloromethyl-p-tolyl sulphide ≥96.0% (by GC)
Chloromethyl-p-tolyl sulphide ≥96.0% (by GC)
TCIAC1601-5G
:
TCI
:
Chloromethyl-p-tolyl sulphide ≥96.0% (by GC)
TCIAC1601-5G
:
TCI
:
Pk:
5 g
Seq:
3173
Appearance:
Colorless to Light yellow clear liquid
Purity (GC):
min. 96.0 %
:
:
172.67 g/mol
:
GHS07
:
Refrigerator
:
34125-84-3
:
C₈H₉ClS
:
MFCD01318451