N-Carbobenzoxy-L-serine-β-lactone ≥98.0% (by HPLC)
TCIAC1575-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Specific rotation [a]20/D:-21.0 to -30.0 deg (C=1, CH3CN)
:
:221.21 g/mol
:Refrigerator
:26054-60-4
:C₁₁H₁₁NO₄
:MFCD01318309