Order Entry
Netherlands
ContactUsLinkComponent
1-Cyclohexyl-2-buten-1-ol (cis and trans mixture) cis- and trans-mixture ≥95.0% (by GC)
1-Cyclohexyl-2-buten-1-ol (cis and trans mixture) cis- and trans-mixture ≥95.0% (by GC)
Catalog # TCIAC1567-5G
Supplier:  TCI
CAS Number:  
1-Cyclohexyl-2-buten-1-ol (cis and trans mixture) cis- and trans-mixture ≥95.0% (by GC)
Catalog # TCIAC1567-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 95.0 % (total of isomers)

Specification Test Results

About this item

 :
 :154.25 g/mol
Boiling Pt:114°C
Flash Pt:66°C
CAS Number:18736-82-8
Formula:C₁₀H₁₈O
MDL Number:MFCD00060819