
Methyl-2-(cyanomethyl)benzoate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:5597-04-6
:
Formula:C₁₀H₉NO₂
MDL Number:MFCD01631304
:175.19 g/mol
Boiling Pt:133°C
UN:3439
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Solubility in Methanol:almost transparency
- Melting point:48.0 to 51.0 deg-C
Specification Test Results
Frequently Bought Together