Order Entry
Netherlands
ContactUsLinkComponent
2,2-Dimethylcyclopropyl cyanide ≥97.0% (by GC)
Flammable
Irritant
2,2-Dimethylcyclopropyl cyanide ≥97.0% (by GC)
Supplier:  TCI
Certificates
Flammable
Irritant
2,2-Dimethylcyclopropyl cyanide ≥97.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

Frequently Bought Together